Hafnium Standard: Hf @ 1000 ug/mL in 2% HNO3/0.5%HF, VHG, LGC Standards
Catalog # 77697-764
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:1000 µg/mL
- Matrix:2% HNO₃/0,5% HF
- Description:Hafnium Standard: Hf @ 1000 ug/mL in 2% HNO3/0.5%HF
- Size:100 ml
- Cat. No.:77697-764