Order Entry
United States
ContactUsLinkComponent
QC Standard 19: As, Be, Ca, Cd, Co, Cr, Cu, Fe, Mg, Mn, Mo, Ni, Pb, Sb, Se, Ti, Tl, V, Zn @ 100 µg/mL in 5% HNO3, tr. F-, tr. Tartaric Acid, VHG, LGC Standards
QC Standard 19: As, Be, Ca, Cd, Co, Cr, Cu, Fe, Mg, Mn, Mo, Ni, Pb, Sb, Se, Ti, Tl, V, Zn @ 100 µg/mL in 5% HNO3, tr. F-, tr. Tartaric Acid, VHG, LGC Standards
Catalog # 77697-839
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
QC Standard 19: As, Be, Ca, Cd, Co, Cr, Cu, Fe, Mg, Mn, Mo, Ni, Pb, Sb, Se, Ti, Tl, V, Zn @ 100 µg/mL in 5% HNO3, tr. F-, tr. Tartaric Acid, VHG, LGC Standards
Catalog # 77697-839
Supplier:  LGC Standards
Supplier Number:  VHG-QC19-100
New Product
CAS Number:  

Specifications

  • Concentration:
    100 µg/mL
  • Matrix:
    5% HNO₃/tr. HF/tr. C₄H₆O₆
  • Description:
    QC Standard 19: As, Be, Ca, Cd, Co, Cr, Cu, Fe, Mg, Mn, Mo, Ni, Pb, Sb, Se, Ti, Tl, V, Zn @ 100 µg/mL in 5% HNO3, tr. F-, tr. Tartaric Acid
  • Size:
    100 ml
  • Cat. No.:
    77697-839

Specifications

About this item