Order Entry
United States
ContactUsLinkComponent
MISA Standard 6: Ag, Al, B, Ba, Ca, Cd, Co, Cr, Cs, Cu, Ga, In, Fe, K, Li, Mg, Mn, Na, Ni, P, Pb, Rb, Sr, Tl, U, V, Zn @ 100 µg/mL in 2% HNO3, VHG, LGC Standards
MISA Standard 6: Ag, Al, B, Ba, Ca, Cd, Co, Cr, Cs, Cu, Ga, In, Fe, K, Li, Mg, Mn, Na, Ni, P, Pb, Rb, Sr, Tl, U, V, Zn @ 100 µg/mL in 2% HNO3, VHG, LGC Standards
Catalog # 77697-693
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
MISA Standard 6: Ag, Al, B, Ba, Ca, Cd, Co, Cr, Cs, Cu, Ga, In, Fe, K, Li, Mg, Mn, Na, Ni, P, Pb, Rb, Sr, Tl, U, V, Zn @ 100 µg/mL in 2% HNO3, VHG, LGC Standards
Catalog # 77697-693
Supplier:  LGC Standards
Supplier Number:  VHG-MISA6-100
New Product
CAS Number:  

Specifications

  • Concentration:
    100 µg/mL
  • Matrix:
    2% HNO₃
  • Description:
    MISA Standard 6: Ag, Al, B, Ba, Ca, Cd, Co, Cr, Cs, Cu, Ga, In, Fe, K, Li, Mg, Mn, Na, Ni, P, Pb, Rb, Sr, Tl, U, V, Zn @ 100 µg/mL in 2% HNO3
  • Size:
    100 ml
  • Cat. No.:
    77697-693

Specifications

About this item