MISA Standard 5: Ce, Dy, Er, Eu, Gd, Ho, La, Lu, Nd, Pr, Sc, Sm, Tb, Th, Tm, Y, Yb @ 100 µg/mL in 2% HNO3, VHG, LGC Standards
Catalog # 77697-692
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:100 µg/mL
- Matrix:2% HNO₃
- Description:MISA Standard 5: Ce, Dy, Er, Eu, Gd, Ho, La, Lu, Nd, Pr, Sc, Sm, Tb, Th, Tm, Y, Yb @ 100 µg/mL in 2% HNO3
- Size:100 ml
- Cat. No.:77697-692