Order Entry
United States
ContactUsLinkComponent
Neodymium Standard: Nd @ 10000 µg/mL in 5% HNO3, VHG, LGC Standards
Neodymium Standard: Nd @ 10000 µg/mL in 5% HNO3, VHG, LGC Standards
Catalog # 77697-900
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Neodymium Standard: Nd @ 10000 µg/mL in 5% HNO3, VHG, LGC Standards
Catalog # 77697-900
Supplier:  LGC Standards
Supplier Number:  VHG-TNDN-250
New Product
CAS Number:  

Specifications

  • Concentration:
    10000 µg/mL
  • Matrix:
    5% HNO₃
  • Description:
    Neodymium Standard: Nd @ 10000 µg/mL in 5% HNO3
  • Size:
    250 ml
  • Cat. No.:
    77697-900

Specifications

About this item