Order Entry
United States
ContactUsLinkComponent
Lead Standard: Pb @ 0.300 g/gal in Isooctane, VHG, LGC Standards
Lead Standard: Pb @ 0.300 g/gal in Isooctane, VHG, LGC Standards
Catalog # 77697-741
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Lead Standard: Pb @ 0.300 g/gal in Isooctane, VHG, LGC Standards
Catalog # 77697-741
Supplier:  LGC Standards
Supplier Number:  VHG-PBISO-0.300-100
New Product
CAS Number:  

Specifications

  • Concentration:
    0.300 g/gal
  • Description:
    Lead Standard: Pb @ 0.300 g/gal in Isooctane
  • Size:
    100 ml
  • Cat. No.:
    77697-741

Specifications

About this item