Rhodium Standard: Rh @ 10 µg/mL in 2% HCl, VHG, LGC Standards
Catalog # 77697-666
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:10 µg/mL
- Matrix:2% HCl
- Description:Rhodium Standard: Rh @ 10 µg/mL in 2% HCl
- Size:100 ml
- Cat. No.:77697-666