Molybdenum Standard: Mo @ 10 µg/mL in 2% HNO3, tr. HF, VHG, LGC Standards
Catalog # 77697-641
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:10 µg/mL
- Matrix:2% HNO₃
- Description:Molybdenum Standard: Mo @ 10 µg/mL in 2% HNO3, tr. HF
- Size:100 ml
- Cat. No.:77697-641