Order Entry
United States
ContactUsLinkComponent
Lead Isotopic Standard: Pb @ 100 µg/mL in 2% HNO3 (Natural abundance), VHG, LGC Standards
Lead Isotopic Standard: Pb @ 100 µg/mL in 2% HNO3 (Natural abundance), VHG, LGC Standards
Catalog # 77697-637
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Lead Isotopic Standard: Pb @ 100 µg/mL in 2% HNO3 (Natural abundance), VHG, LGC Standards
Catalog # 77697-637
Supplier:  LGC Standards
Supplier Number:  VHG-LISPB1-50
New Product
CAS Number:  

Specifications

  • Concentration:
    100 µg/mL
  • Matrix:
    2% HNO₃
  • Description:
    Lead Isotopic Standard: Pb @ 100 µg/mL in 2% HNO3 (Natural abundance)
  • Size:
    50 ml
  • Cat. No.:
    77697-637

Specifications

About this item