Multi-Anion Standard 1: F-, Cl-, Br-, NO3-, PO4(-3), SO4(-2) @ 100 µg/mL in H2O, VHG, LGC Standards
Catalog # 77697-543
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:100 µg/mL
- Matrix:H₂O
- Description:Multi-Anion Standard 1: F-, Cl-, Br-, NO3-, PO4(-3), SO4(-2) @ 100 µg/mL in H2O
- Size:100 ml
- Cat. No.:77697-543