Order Entry
United States
ContactUsLinkComponent
ICH/USP Parenteral Combined-1 Target Elements: Mo @ 1500, Cr @ 1100, Ba @ 700, Sn @ 600, Cu @ 300, Li @ 250, Sb @ 90, Se @ 80, Ag @ 10, Tl @ 8 µg/mL in 5% HNO3, tr. HF, VHG, LGC Standards
ICH/USP Parenteral Combined-1 Target Elements: Mo @ 1500, Cr @ 1100, Ba @ 700, Sn @ 600, Cu @ 300, Li @ 250, Sb @ 90, Se @ 80, Ag @ 10, Tl @ 8 µg/mL in 5% HNO3, tr. HF, VHG, LGC Standards
Catalog # 77697-540
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
ICH/USP Parenteral Combined-1 Target Elements: Mo @ 1500, Cr @ 1100, Ba @ 700, Sn @ 600, Cu @ 300, Li @ 250, Sb @ 90, Se @ 80, Ag @ 10, Tl @ 8 µg/mL in 5% HNO3, tr. HF, VHG, LGC Standards
Catalog # 77697-540
Supplier:  LGC Standards
Supplier Number:  VHG-ICH-USP-TELF-100
New Product
CAS Number:  

Specifications

  • Concentration:
    Varies
  • Matrix:
    5% HNO₃/tr HF
  • Description:
    ICH/USP Parenteral Combined-1 Target Elements: Mo @ 1500, Cr @ 1100, Ba @ 700, Sn @ 600, Cu @ 300, Li @ 250, Sb @ 90, Se @ 80, Ag @ 10, Tl @ 8 µg/mL in 5% HNO3, tr. HF
  • Size:
    100 ml
  • Cat. No.:
    77697-540

Specifications

About this item