ICH/USP Oral Target Elements Standard C: Au, Ir, Os, Pd, Pt, Rh, Ru @ 100 µg/mL in 15% HCl, VHG, LGC Standards
Catalog # 77697-536
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:100 µg/mL
- Matrix:15% HCl
- Description:ICH/USP Oral Target Elements Standard C: Au, Ir, Os, Pd, Pt, Rh, Ru @ 100 µg/mL in 15% HCl
- Size:100 ml
- Cat. No.:77697-536