ICP-OES Instrument Calibration Standard 4: As, Tl @ 100 µg/mL; Cd, Pb, Se @ 50 µg/mL in 5% HNO3, VHG, LGC Standards
Catalog # 77697-599
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:Varies
- Matrix:5% HNO₃
- Description:ICP-OES Instrument Calibration Standard 4: As, Tl @ 100 µg/mL; Cd, Pb, Se @ 50 µg/mL in 5% HNO3
- Size:100 ml
- Cat. No.:77697-599