Order Entry
United States
ContactUsLinkComponent
 
Sodium Standard: Na+ @ 100 µg/mL in H2O, VHG, LGC Standards
  77697-573
New Product
 :  
undefined
Sodium Standard: Na+ @ 100 µg/mL in H2O, VHG, LGC Standards
  77697-573
 :  VHG-INAW100-500
New Product
 :  

 

  • Concentration:
    100 µg/mL
  • Matrix:
    H₂O
  • Description:
    Sodium Standard: Na+ @ 100 µg/mL in H2O
  • Size:
    500 ml
  • Cat. No.:
    77697-573