Order Entry
United States
ContactUsLinkComponent
 
Magnesium Standard: Mg @ 1000 µg/mL in 5% HNO3, VHG, LGC Standards
  77697-781
New Product
 :  
undefined
Magnesium Standard: Mg @ 1000 µg/mL in 5% HNO3, VHG, LGC Standards
  77697-781
 :  VHG-PMGN-500
New Product
 :  

 

  • Concentration:
    1000 µg/mL
  • Matrix:
    5% HNO₃
  • Description:
    Magnesium Standard: Mg @ 1000 µg/mL in 5% HNO3
  • Size:
    500 ml
  • Cat. No.:
    77697-781