Fluoride Ion Chromatography Standard: F- @ 100µg/mL in H2O, VHG, LGC Standards
Supplier: LGC Standards
New Product
Certificates
Fluoride Ion Chromatography Standard: F- @ 100µg/mL in H2O, VHG, LGC Standards
Supplier: LGC Standards
New Product
About this item
Frequently Bought Together
Screw Top Vials and Caps, 2 ml, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
Dulbecco's phosphate-buffered saline (DPBS), Mediatech
Supplier: Corning
Ward's® Prepared Potato Dextrose Agar
Supplier: Culture Media & Supplies
VWR® Razor Blades
Supplier: VWR International
GelRed® & GelGreen® Nucleic Acid Gel Stains, Biotium
Supplier: Biotium
Corning® DMEM (Dulbecco's Modification of Eagle's Medium), Corning
Supplier: Corning
Upchurch Scientific® Flat-Bottom Fittings, For Tubing Ø > ¹/₁₆" (1.6 mm) and ≤ ¹/₈" (3.2 mm), IDEX Health & Science
Supplier: Upchurch Scientific
Vaplock™ Chemical Exhaust Filters and Color Indicators, Cole-Parmer
Supplier: Antyila Scientific
VWR® Syringe Filters
Supplier: VWR International
VWR® Redi-Stor™ Conductivity Probe Storage Solution
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
Tissue-Tek® O.C.T. Compound, Sakura® Finetek
Supplier: Sakura Finetek
Corning® PBS (Phosphate-Buffered Saline), Corning
Supplier: Corning
VWR® Chemical-Resistant Laboratory Marker
Supplier: VWR International
Blood Agar Plate, 5% Sheep Blood in Tryptic Soy Agar (TSA) Base, 15×100 mm plate
Supplier: Hardy Diagnostics
Flow Cells for MinION™ and GridION™ Devices
Supplier: Oxford Nanopore Technologies