Order Entry
United States
ContactUsLinkComponent
Multi-Anion Standard 4: F- @ 100; Cl- @ 200; Br-, NO3-, SO4(-2) @ 400; PO4(-3) @ 600 µg/mL in H2O, VHG, LGC Standards
Multi-Anion Standard 4: F- @ 100; Cl- @ 200; Br-, NO3-, SO4(-2) @ 400; PO4(-3) @ 600 µg/mL in H2O, VHG, LGC Standards
Catalog # 77697-546
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Multi-Anion Standard 4: F- @ 100; Cl- @ 200; Br-, NO3-, SO4(-2) @ 400; PO4(-3) @ 600 µg/mL in H2O, VHG, LGC Standards
Catalog # 77697-546
Supplier:  LGC Standards
Supplier Number:  VHG-ICM4-100
New Product
CAS Number:  

Specifications

  • Concentration:
    Varies
  • Matrix:
    H₂O
  • Description:
    Multi-Anion Standard 4: F- @ 100; Cl- @ 200; Br-, NO3-, SO4(-2) @ 400; PO4(-3) @ 600 µg/mL in H2O
  • Size:
    100 ml
  • Cat. No.:
    77697-546

Specifications

About this item