Multi-Anion Standard 3: F- @ 20; Cl- @ 30; NO3- @ 100; PO4(-3), SO4(-2) @ 150 µg/mL in H2O, VHG, LGC Standards
Catalog # 77697-545
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:Varies
- Matrix:H₂O
- Description:Multi-Anion Standard 3: F- @ 20; Cl- @ 30; NO3- @ 100; PO4(-3), SO4(-2) @ 150 µg/mL in H2O
- Size:100 ml
- Cat. No.:77697-545