Order Entry
United States
ContactUsLinkComponent
ICP-AES CRQL Solution 2: Ca, K, Mg, Na @ 5000 µg/mL in 5% HNO3, VHG, LGC Standards
ICP-AES CRQL Solution 2: Ca, K, Mg, Na @ 5000 µg/mL in 5% HNO3, VHG, LGC Standards
Catalog # 77697-480
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
ICP-AES CRQL Solution 2: Ca, K, Mg, Na @ 5000 µg/mL in 5% HNO3, VHG, LGC Standards
Catalog # 77697-480
Supplier:  LGC Standards
Supplier Number:  VHG-CRQL2AES-100
New Product
CAS Number:  

Specifications

  • Concentration:
    5000 µg/mL
  • Matrix:
    5% HNO₃
  • Description:
    ICP-AES CRQL Solution 2: Ca, K, Mg, Na @ 5000 µg/mL in 5% HNO3
  • Size:
    100 ml
  • Cat. No.:
    77697-480

Specifications

About this item