Order Entry
United States
ContactUsLinkComponent
Rhodium Standard: Rh @ 1000 µg/mL in 20% HCl, VHG, LGC Standards
Rhodium Standard: Rh @ 1000 µg/mL in 20% HCl, VHG, LGC Standards
Catalog # 77697-814
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Rhodium Standard: Rh @ 1000 µg/mL in 20% HCl, VHG, LGC Standards
Catalog # 77697-814
Supplier:  LGC Standards
Supplier Number:  VHG-PRHH-100
New Product
CAS Number:  

Specifications

  • Concentration:
    1000 µg/mL
  • Matrix:
    20% HCl
  • Description:
    Rhodium Standard: Rh @ 1000 µg/mL in 20% HCl
  • Size:
    100 ml
  • Cat. No.:
    77697-814

Specifications

About this item