ICP-MS Internal Standard Solution: Bi, Ge, In, Li-6, Rh, Sc, Tb, Y @ 100ug/mL in 5% HNO3, tr. F-, VHG, LGC Standards
77697-628
New Product
:
- Concentration:100 µg/mL
- Matrix:5% HNO₃/tr HF
- Description:ICP-MS Internal Standard Solution: Bi, Ge, In, Li-6, Rh, Sc, Tb, Y @ 100ug/mL in 5% HNO3, tr. F-
- Size:100 ml
- Cat. No.:77697-628