Selenium Isotopic Standard: Se @ 10 µg/mL in 2% HNO3 (Enriched in Se-78), VHG, LGC Standards
Catalog # 77697-627
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:10 µg/mL
- Matrix:2% HNO₃
- Description:Selenium Isotopic Standard: Se @ 10 µg/mL in 2% HNO3 (Enriched in Se-78)
- Size:50 ml
- Cat. No.:77697-627