Internal Standard Multi-Element Mix 7: Te @ 25; Sc @ 10; Ge, In, Lu, Bi @ 5 µg/mL in 2% HNO3, tr. HF, VHG, LGC Standards
Catalog # 77697-626
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:Varies
- Matrix:2% HNO₃/tr. HF
- Description:Internal Standard Multi-Element Mix 7: Te @ 25; Sc @ 10; Ge, In, Lu, Bi @ 5 µg/mL in 2% HNO3, tr. HF
- Size:100 ml
- Cat. No.:77697-626