Order Entry
United States
ContactUsLinkComponent
Nickel Isotopic Standard: Ni @ 10 µg/mL in 2% HNO3 (Enriched in Ni-61), VHG, LGC Standards
Nickel Isotopic Standard: Ni @ 10 µg/mL in 2% HNO3 (Enriched in Ni-61), VHG, LGC Standards
Catalog # 77697-624
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Nickel Isotopic Standard: Ni @ 10 µg/mL in 2% HNO3 (Enriched in Ni-61), VHG, LGC Standards
Catalog # 77697-624
Supplier:  LGC Standards
Supplier Number:  VHG-LIS61NI-50
New Product
CAS Number:  

Specifications

  • Concentration:
    10 µg/mL
  • Matrix:
    2% HNO₃
  • Description:
    Nickel Isotopic Standard: Ni @ 10 µg/mL in 2% HNO3 (Enriched in Ni-61)
  • Size:
    50 ml
  • Cat. No.:
    77697-624

Specifications

About this item