Order Entry
United States
ContactUsLinkComponent
Pharma Metals Calibration Standard 1: B, Ba, Cu, Fe, Zn @ 100; Ag, Al, As, Cd, Co, Cr, Li, Mn, Mo, Ni, Pb, Sb, Se, Sn, Tl, V @ 20 µg/mL in 2% HNO3/tr. HF, VHG, LGC Standards
Pharma Metals Calibration Standard 1: B, Ba, Cu, Fe, Zn @ 100; Ag, Al, As, Cd, Co, Cr, Li, Mn, Mo, Ni, Pb, Sb, Se, Sn, Tl, V @ 20 µg/mL in 2% HNO3/tr. HF, VHG, LGC Standards
Catalog # 77697-762
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Pharma Metals Calibration Standard 1: B, Ba, Cu, Fe, Zn @ 100; Ag, Al, As, Cd, Co, Cr, Li, Mn, Mo, Ni, Pb, Sb, Se, Sn, Tl, V @ 20 µg/mL in 2% HNO3/tr. HF, VHG, LGC Standards
Catalog # 77697-762
Supplier:  LGC Standards
Supplier Number:  VHG-PHARM1-100
New Product
CAS Number:  

Specifications

  • Concentration:
    Varies
  • Description:
    Pharma Metals Calibration Standard 1: B, Ba, Cu, Fe, Zn @ 100; Ag, Al, As, Cd, Co, Cr, Li, Mn, Mo, Ni, Pb, Sb, Se, Sn, Tl, V @ 20 µg/mL in 2% HNO3/tr. HF
  • Size:
    100 ml
  • Cat. No.:
    77697-762

Specifications

About this item