Scandium Standard: Sc @ 1000 µg/g in Hydrocarbon Oil, VHG, LGC Standards
Catalog # 77697-727
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:1000 µg/mL
- Matrix:oil
- Description:Scandium Standard: Sc @ 1000 µg/g in Hydrocarbon Oil
- Size:50 ml
- Cat. No.:77697-727