Lubricating Oil Standard 7: Ca @ 0.200; P @ 0.080; S @ 0.075; Zn @ 0.120 wt% in Lubricating Oil, VHG, LGC Standards
Catalog # 77697-660
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:Varies
- Matrix:oil
- Description:Lubricating Oil Standard 7: Ca @ 0.200; P @ 0.080; S @ 0.075; Zn @ 0.120 wt% in Lubricating Oil
- Size:100 ml
- Cat. No.:77697-660