Lubricating Oil Standard 4: Ca @ 0.500; P @ 0.030 S @ 0.150; Zn @ 0.070 wt% in Lubricating Oil, VHG, LGC Standards
77697-657
New Product
:
- Concentration:Varies
- Matrix:oil
- Description:Lubricating Oil Standard 4: Ca @ 0.500; P @ 0.030 S @ 0.150; Zn @ 0.070 wt% in Lubricating Oil
- Size:100 ml
- Cat. No.:77697-657