Lubricating Oil Standard 3: Ca @ 0.000; P @ 0.020; S @ 0.300; Zn @ 0.175 wt% in Lubricating Oil, VHG, LGC Standards
Catalog # 77697-656
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:Varies
- Matrix:oil
- Description:Lubricating Oil Standard 3: Ca @ 0.000; P @ 0.020; S @ 0.300; Zn @ 0.175 wt% in Lubricating Oil
- Size:100 ml
- Cat. No.:77697-656