Lubricating Oil Standard 2: Ca @ 0.600; P @ 0.005; S @ 0.050; Zn @ 0.080 wt% in Lubricating Oil, VHG, LGC Standards
77697-655
New Product
:
- Concentration:Varies
- Matrix:oil
- Description:Lubricating Oil Standard 2: Ca @ 0.600; P @ 0.005; S @ 0.050; Zn @ 0.080 wt% in Lubricating Oil
- Size:100 ml
- Cat. No.:77697-655