Lubricating Oil Standard 15: Ca @ 0.150; P @ 0.100; S @ 0.250; Zn @ 0.090 wt% in Lubricating Oil, VHG, LGC Standards
77697-653
New Product
:
- Concentration:Varies
- Matrix:oil
- Description:Lubricating Oil Standard 15: Ca @ 0.150; P @ 0.100; S @ 0.250; Zn @ 0.090 wt% in Lubricating Oil
- Size:100 ml
- Cat. No.:77697-653