Lubricating Oil Standard 1: Ca @ 0.000; P @ 0.000; S @ 0.000; Zn @ 0.000 wt% in Lubricating Oil, VHG, LGC Standards
77697-648
New Product
:
- Concentration:Varies
- Matrix:oil
- Description:Lubricating Oil Standard 1: Ca @ 0.000; P @ 0.000; S @ 0.000; Zn @ 0.000 wt% in Lubricating Oil
- Size:100 ml
- Cat. No.:77697-648