Nickel Standard: Ni @ 10 µg/mL in 2% HNO3, VHG, LGC Standards
Catalog # 77697-646
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:10 µg/mL
- Matrix:2% HNO₃
- Description:Nickel Standard: Ni @ 10 µg/mL in 2% HNO3
- Size:100 ml
- Cat. No.:77697-646