ICP-MS Tuning Solution: Be, Mg, Co, In, Ba, Ce, Tl, Pb, Th @ 250 ug/L in 2% HNO3, VHG, LGC Standards
Catalog # 77697-645
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:250 µg/L
- Matrix:2% HNO₃
- Description:ICP-MS Tuning Solution: Be, Mg, Co, In, Ba, Ce, Tl, Pb, Th @ 250 ug/L in 2% HNO3
- Size:500 ml
- Cat. No.:77697-645