Order Entry
United States
ContactUsLinkComponent
ICP-MS Tuning Solution: 7Li, Co, Y, Ce, Tl @ 10 ug/L in 2% HNO3, VHG, LGC Standards
ICP-MS Tuning Solution: 7Li, Co, Y, Ce, Tl @ 10 ug/L in 2% HNO3, VHG, LGC Standards
Catalog # 77697-644
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
ICP-MS Tuning Solution: 7Li, Co, Y, Ce, Tl @ 10 ug/L in 2% HNO3, VHG, LGC Standards
Catalog # 77697-644
Supplier:  LGC Standards
Supplier Number:  VHG-LMSTNG5DIL-500
New Product
CAS Number:  

Specifications

  • Concentration:
    10 µg/mL
  • Matrix:
    2% HNO₃
  • Description:
    ICP-MS Tuning Solution: 7Li, Co, Y, Ce, Tl @ 10 ug/L in 2% HNO3
  • Size:
    500 ml
  • Cat. No.:
    77697-644

Specifications

About this item