Internal Standard Multi-Element Mix 2: Ga, Y, In, Tb, Bi @ 20; 6Li, Sc @ 100 µg/mL in 2% HNO3, VHG, LGC Standards
77697-621
New Product
:
- Concentration:Varies
- Matrix:2% HNO₃
- Description:Internal Standard Multi-Element Mix 2: Ga, Y, In, Tb, Bi @ 20; 6Li, Sc @ 100 µg/mL in 2% HNO3
- Size:100 ml
- Cat. No.:77697-621