Internal Standard Stock Solution for Methods 200.8 & CLP ILM05.2: 6Li, Sc, Y, In, Tb, Lu, Bi @ 10 µg/mL in 5% HNO3, VHG, LGC Standards
Catalog # 77697-620
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:10 µg/mL
- Matrix:5% HNO₃
- Description:Internal Standard Stock Solution for Methods 200.8 & CLP ILM05.2: 6Li, Sc, Y, In, Tb, Lu, Bi @ 10 µg/mL in 5% HNO3
- Size:100 ml
- Cat. No.:77697-620