Environmental ICV Std A (Second Source) 26 Component Standard in 5% HNO3, tr. HF, tr. Tartaric Acid, VHG, LGC Standards
Catalog # 77697-617
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:Varies
- Matrix:5% HNO₃/tr HF
- Description:Environmental ICV Std A (Second Source): Ag, Al, As, Ba, Be, Cd, Co, Cr, Cu, Mn, Mo, Ni, Pb, Sb, Se, Sr, Th, Tl, U, V, Zn @ 10; Ca, Fe, K, Mg, Na @ 1000 µg/mL in 5% HNO3, tr. HF, tr. Tartaric Acid
- Size:100 ml
- Cat. No.:77697-617