Order Entry
United States
ContactUsLinkComponent
Mercury Standard: Hg @ 10 µg/mL in 5% HNO3, VHG, LGC Standards
Mercury Standard: Hg @ 10 µg/mL in 5% HNO3, VHG, LGC Standards
Catalog # 77697-616
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Mercury Standard: Hg @ 10 µg/mL in 5% HNO3, VHG, LGC Standards
Catalog # 77697-616
Supplier:  LGC Standards
Supplier Number:  VHG-LHGN-100
New Product
CAS Number:  

Specifications

  • Concentration:
    10 µg/mL
  • Matrix:
    5% HNO₃
  • Description:
    Mercury Standard: Hg @ 10 µg/mL in 5% HNO3
  • Size:
    100 ml
  • Cat. No.:
    77697-616

Specifications

About this item