Iron AA Standard: Fe @ 1000 µg/mL in 5% HNO3, VHG, LGC Standards
Supplier: LGC Standards
New Product
Certificates
About this item
Frequently Bought Together
Whatman™ GD/XP Syringe Filters
Supplier: Cytiva
Whatman™ Anotop Syringe Filters, Non Sterile
Supplier: Cytiva
Whatman™ Grade 5 Qualitative Filter Papers
Supplier: Cytiva
J.T.Baker®, Syringe Filters
Supplier: Avantor
700 Series MICROLITER® Syringes, Hamilton
Supplier: Hamilton
Titan3™ PTFE (Hydrophobic) Syringe Filters, Thermo Scientific
Supplier: Thermo Scientific
Acclaim™ 120 C8 LC Columns, Thermo Scientific
Supplier: Thermo Scientific
MICRO 20CM 0.65UM MPES 0.75MM
Supplier: 0000019223
FILTER SYS 150ML PES 0.45UM 50MM S CS12
Supplier: CELLTREAT SCIENTIFIC PRODUCTS MS
SunFlo™ Dissolution Filters, SUN-SRi
Supplier: Thermo Scientific
FILTER SYS 1000ML PES 0.22UM 90MM S CS12
Supplier: VistaLab
MICRO 20CM 0.2UM PES MLLXFLL 0.5MM
Supplier: 0000019223
Minisart® RC Syringe Filters
Supplier: Sartorius
Minisart® Syringe Filters NML
Supplier: Sartorius