Lead in Paint Chips Reference Material: Pb approx. 643 µg/g, VHG, LGC Standards
Catalog # 77697-748
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:643 µg/g
- Description:Lead in Paint Chips Reference Material: Pb approx. 643 µg/g
- Size:50 ml
- Cat. No.:77697-748