Order Entry
United States
ContactUsLinkComponent
ICP-MS Tuning Solution 1: Ce, Co, Li, Tl, Y @ 10 ug/mL in 2% HNO3, VHG, LGC Standards
ICP-MS Tuning Solution 1: Ce, Co, Li, Tl, Y @ 10 ug/mL in 2% HNO3, VHG, LGC Standards
Catalog # 77697-610
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
ICP-MS Tuning Solution 1: Ce, Co, Li, Tl, Y @ 10 ug/mL in 2% HNO3, VHG, LGC Standards
Catalog # 77697-610
Supplier:  LGC Standards
Supplier Number:  VHG-LAGTSTK1-100
New Product
CAS Number:  

Specifications

  • Concentration:
    10 µg/mL
  • Matrix:
    2% HNO₃
  • Description:
    ICP-MS Tuning Solution : Ce, Co, Li, Tl, Y @ 10 ug/mL in 2% HNO3
  • Size:
    100 ml
  • Cat. No.:
    77697-610

Specifications

About this item