Sn @ 5µg/mL; Si @ 10µg/mL; Mo @ 20µg/mL in 3% HNO3 (v/v)/tr HF, VHG, LGC Standards
Catalog # 77697-444
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:Varies
- Description:Sn @ 5µg/mL; Si @ 10µg/mL; Mo @ 20µg/mL in 3% HNO3 (v/v)/tr HF, 500mL
- Size:500 ml
- Cat. No.:77697-444