Order Entry
United States
ContactUsLinkComponent
Manganese AA Standard: Mn @ 1000 µg/mL in 5% HNO3, VHG, LGC Standards
Manganese AA Standard: Mn @ 1000 µg/mL in 5% HNO3, VHG, LGC Standards
Catalog # 77697-464
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Manganese AA Standard: Mn @ 1000 µg/mL in 5% HNO3, VHG, LGC Standards
Catalog # 77697-464
Supplier:  LGC Standards
Supplier Number:  VHG-AAMNN-100
New Product
CAS Number:  

Specifications

  • Concentration:
    1000 µg/mL
  • Matrix:
    5% HNO₃
  • Description:
    Manganese AA Standard: Mn @ 1000 µg/mL in 5% HNO3
  • Size:
    100 ml
  • Cat. No.:
    77697-464

Specifications

About this item