$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
United States
ContactUsLinkComponent
Standards
Organic Standards
Acetaldehyde, TRC, LGC Standards
Acetaldehyde, TRC, LGC Standards
77693-745
:
LGC Standards
New Product
:
Acetaldehyde, TRC, LGC Standards
77693-745
:
LGC Standards
:
TRC-A132600-1G
New Product
:
CAS No.:
75-07-0
Description:
Acetaldehyde
Size:
1 g
Formula:
CH₃CHO
Molecular Weight:
44.05 g/mol
Cat. No.:
77693-745