Sertaconazole Nitrate, Mikromol, LGC Standards
Catalog # 77697-408
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:99592-39-9
- Description:Sertaconazole Nitrate
- Size:250 mg
- Formula:C₂₀H₁₅Cl₃N₂OS·HNO₃
- Molecular Weight:500.79 g/mol
- Cat. No.:77697-408