Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Homosalate, Mikromol, LGC Standards
Homosalate, Mikromol, LGC Standards
Homosalate, Mikromol, LGC Standards
77697-402
:
LGC Standards
New Product
:
Homosalate, Mikromol, LGC Standards
77697-402
:
LGC Standards
:
MM1570.00
New Product
:
CAS No.:
118-56-9
Description:
Homosalate
Size:
250 mg
Formula:
C₁₆H₂₂O₃
Molecular Weight:
262.35 g/mol
Cat. No.:
77697-402