Iopamidol, Mikromol, LGC Standards
Catalog # 77697-355
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:60166-93-0
- Description:Iopamidol
- Size:250 mg
- Formula:C₁₇H₂₂I₃N₃O₈
- Molecular Weight:777.09 g/mol
- Cat. No.:77697-355