$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
United States
ContactUsLinkComponent
Standards
Organic Standards
p-Chloroamphetamine Hydrochloride, LoGiCal, LGC Standards
p-Chloroamphetamine Hydrochloride, LoGiCal, LGC Standards
77697-147
:
LGC Standards
New Product
:
p-Chloroamphetamine Hydrochloride, LoGiCal, LGC Standards
77697-147
:
LGC Standards
:
LGCFOR1388.04
New Product
:
CAS No.:
3706-38-5
Description:
p-Chloroamphetamine Hydrochloride
Size:
10 mg
Cat. No.:
77697-147