O-Desmethylvenlafaxine, LoGiCal, LGC Standards
Catalog # 77697-091
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:93413-62-8
- Description:O-Desmethylvenlafaxine
- Size:10 mg
- Formula:C₁₆H₂₅NO₂
- Molecular Weight:263.38 g/mol
- Cat. No.:77697-091